Learn More
Hemoglobin beta C Monoclonal Antibody (15C2.C11.F2.G11), Rockland™

Mouse Monoclonal Antibody
$174.73 - $651.91
Specifications
Antigen | Hemoglobin beta C |
---|---|
Clone | 15C2.C11.F2.G11 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0625
![]() ![]() |
Rockland Immunochemicals
200301GS7 |
100 μg |
Each for $651.91
|
|
|||||
50-199-0626
![]() ![]() |
Rockland Immunochemicals
200301GS7S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. This product is stable at 4°C as an undiluted liquid. For extended storage, aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Dilute only prior to immediate use. Reacts specifically with human HbC beta c-variant isoform. This antibody does not react with the HbA, HbS, HbF, or HbA-2 isoforms.
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.Specifications
Hemoglobin beta C | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
beta globin chain; Beta-1-globin; Beta-globin; CD113t-C; HBB; Hemoglobin beta chain; hemoglobin beta chain complex; Hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; Hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin | |
HBB | |
IgG1 κ | |
-20°C, Avoid Freeze/Thaw Cycles | |
HBB |
15C2.C11.F2.G11 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P68871 | |
3043 | |
Anti-Hemoglobin beta C Monoclonal Antibody was produced in mice by repeated immunizations with synthetic peptide corresponding to amino acid residues near the N-terminus of Hb beta-subunit conjugated to KLH. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.