Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin beta S Monoclonal Antibody (23E5.H6.G6.C1.H7.F7.G9.F6), Rockland™
SDP

Catalog No. 501990621
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0621 100 μg
50-199-0622 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0621 Supplier Rockland Immunochemicals Supplier No. 200301GS5
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. This product is stable at 4°C as an undiluted liquid. For extended storage, aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Dilute only prior to immediate use. Reacts specifically with human HbS beta sickle isoform. This antibody does not react with the HbA, HbF, HbC, or HbA-2 isoform.

The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.

Specifications

Antigen Hemoglobin beta S
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23E5.H6.G6.C1.H7.F7.G9.F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene HBB
Gene Accession No. P68871
Gene Alias beta globin chain; Beta-1-globin; Beta-globin; CD113t-C; HBB; Hemoglobin beta chain; hemoglobin beta chain complex; Hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; Hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin
Gene Symbols HBB
Host Species Mouse
Immunogen Anti-Hemoglobin beta S Monoclonal Antibody was produced in mice by repeated immunizations with synthetic peptide corresponding to amino acid residues near the N-terminus of Hb beta-subunit conjugated to KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3043
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.