Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemoglobin gamma-1,2 Recombinant Rabbit Monoclonal Antibody (JM84-10)
GREENER_CHOICE

Catalog No. PIMA532603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532603 Supplier Invitrogen™ Supplier No. MA532603
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregnancy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects. Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology, referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described with increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin gamma-1,2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JM84-10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene HBG1
Gene Accession No. P69891, P69892
Gene Alias abnormal hemoglobin; A-gamma globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Gamma-2-globin; G-gamma globin Paulinia; hb F Agamma; Hb F Ggamma; HBG1; HBG2; HBGA; HBGR; HBG-T1; HBG-T2; Hemoglobin gamma-1 chain; hemoglobin gamma-2 chain; Hemoglobin gamma-A chain; hemoglobin gamma-G chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma 2; hemoglobin subunit gamma-1; Hemoglobin subunit gamma-2; hemoglobin, gamma A; hemoglobin, gamma G; hemoglobin, gamma, regulator of; HSGGL1; methemoglobin; PRO2979; TNCY
Gene Symbols HBG1, HBG2
Host Species Rabbit
Immunogen Synthetic peptide within Human HBG1/2 aa 2-45.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047, 3048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.