Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemoglobin gamma A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577998 Supplier Invitrogen™ Supplier No. PA577998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: K562 Predicted Reactivity: Chimpanzee (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5'-epsilon - gamma-G - gamma-A - delta - beta-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin gamma A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Hbg1
Gene Accession No. P69891
Gene Alias A-gamma globin; epsilon 3 globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Hb F Agamma; Hbb-y; Hbe1; Hbg1; HBGA; HBGR; HBG-T2; Hemoglobin gamma-1 chain; hemoglobin gamma-a chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma-1; hemoglobin Y beta-like embryonic chain; hemoglobin Y, beta-like embryonic chain; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; HSGGL1; PRO2979
Gene Symbols Hbg1
Host Species Rabbit
Immunogen Full length human Hemoglobin gamma A Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047, 94164
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.