Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemoglobin Goat anti-Human, HRP, Polyclonal , Abnova™
Goat polyclonal antibody raised against human Hemoglobin.
Supplier: Abnova Corporation PAB29658
Specifications
Hemoglobin | |
Polyclonal | |
Goat polyclonal antibody raised against human Hemoglobin. | |
In 50mM HEPES (Stabilizer, 0.002% Thimerosal) | |
Native purified human Hemoglobin. | |
1 mg | |
Primary | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Western Blot | |
HRP | |
ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
Goat | |
Antigen Affinity Purification | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction