Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin Mouse anti-Human, Clone: M1709Hg2, Abnova™

Catalog No. 89934233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-233 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-233 Supplier Abnova Corporation Supplier No. MAB12863
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human Hemoglobin.

Specifications

Antigen Hemoglobin
Applications ELISA, Immunohistochemistry (PFA fixed), Lateral Flow
Classification Monoclonal
Clone M1709Hg2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human Hemoglobin.
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, 50mM NaCl, pH 8.0 (0.05% sodium azide).
Host Species Mouse
Immunogen Human Hemoglobin.
Purification Method Ion exchange chromatography purification
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.