Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemopexin Antibody, Janelia Fluor™ 549, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP311033JF549
Description
Hemopexin Polyclonal antibody specifically detects Hemopexin in Human samples. It is validated for ELISASpecifications
Hemopexin | |
Polyclonal | |
ELISA | |
beta-1B-glycoprotein, FLJ56652, hemopexin, HX | |
Human Hemopexin | |
0.1 mL | |
Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
3263 | |
Store at 4°C in the dark. |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Goat | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction