Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemopexin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321321100UL
Description
Hemopexin Polyclonal antibody specifically detects Hemopexin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Hemopexin | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
beta-1B-glycoprotein, FLJ56652, hemopexin, HX | |
This antibody was developed against Recombinant Protein corresponding to amino acids: ECHRGECQAEGVLFFQGDREWFWDLATGTMKERSWPAVGNCSSALRWLGRYYCFQGNQFLRFDPVRGEVPPRYPRDVRDYFMPCP | |
100 μg | |
Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases | |
3263 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction