Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemopexin Monoclonal Antibody (2H4C4F12)
GREENER_CHOICE

Catalog No. PIMA530635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530635 Supplier Invitrogen™ Supplier No. MA530635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human HPX/Hemopexin.

Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemopexin
Applications ELISA
Classification Monoclonal
Clone 2H4C4F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Hpx
Gene Accession No. P02790
Gene Alias Beta-1B-glycoprotein; hemopexin; HPX; Hpxn; hx; hyaluronidase
Gene Symbols Hpx
Host Species Mouse
Immunogen Recombinant Human HPX protein (Met1-His462).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3263
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.