Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemopexin Monoclonal Antibody (32)
GREENER_CHOICE

Catalog No. ABA133202
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABA-133202 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABA-133202 Supplier Invitrogen™ Supplier No. ABS0133202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 013-32-02 detects Hemopexin in human samples. ABS 013-32-02 has been successfully used in ELISA procedures. ABS 013-32-02 binds Hemopexin when coated on ELISA wells or presented by an antibody capture coat of ABS 013-04-02.

Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemopexin
Applications ELISA
Classification Monoclonal
Clone 32
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Hpx
Gene Accession No. P02790
Gene Alias Beta-1B-glycoprotein; hemopexin; HPX; Hpxn; hx; hyaluronidase
Gene Symbols Hpx
Host Species Mouse
Immunogen Hemopexin purified from human plasma.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3263
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.