Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hemosure™ iFOB Test Kits

Click to view available options
Includes:
10 tests, 10 tubes, 10 mailers
20 tests, 10 home mailers
20 tests, 20 home mailers
30 tests, 30 tubes, no mailers
30 tests, 50 tubes, 50 mailers
50 tests, 50 tubes, no mailers
Description
- CLIA Complexity: Waived
- CPT Codes: G0328QW (colorectal cancer screening); 82274QW (detection of blood in feces form)
- Sensitivity: 50ng hHB/mL
- Specificity: over 96%
- Useful aid in the diagnosis and therapy of gastrointestinal disorders
- Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes
- As test sample flows up through absorbent device, the labeled antibody-dye conjugate binds to the hemoglobin in specimen, forming an antibody-antigen complex
- Results within five minutes
- Positive results: one band in the control (C) region, one band in the test (T) region
- Negative results: only one band in the control (C) region
- Low false positive results
- Reaction components: citric acid, monoclonal and polyclonal antibody, mouse anti-HB antibodies 1 and 2, and goat anti-mouse IgG antibody
If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.
Order Info
The 10-test kit contains 10 collection bottles and 10 home kit mailers; 30-test kit contains 50 collection bottles and 50 home kit mailers.
Specifications
Specifications
| Content And Storage | 20 Fecal occult Blood test cassettes, 20 home kit mailer kits (each kit includes 1 collection tube, 2 collection Papers, patient instructions and return envelope) |
| CPT Code | G0328QW, 82274QW |
| Clia Complexity | Waived |
| Type | Pre-pack iFOB20 |
| Includes | 20 tests, 20 home mailers |
| Quantity | 20 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction