Learn More
Delcon HemoWeld-Gun Heavy Duty Portable Sealer
Catalog No. 22004213
Change view
Click to view available options
Includes:
Without Transport Case
Catalog No. Includes
22-004-213 Without Transport Case
1 options
This item is not returnable. View return policy
Catalog No. 22-004-213 Supplier Delcon Supplier No. HWG2.00
Add to Cart
Add to Cart
This item is not returnable. View return policy
HemoWeld-Gun heavy duty portable sealer is an innovative sealer with exclusive tube retention system that minimizes errors to achieve perfect seals. Features include ergonomic design, durable silicone cable and long-lasting battery.
  • RF generator in handle eliminates power loss from cord
  • Power status indicator is provided after each seal
  • Strong and durable seal
  • Heavy duty motor head
  • 1500 seals per charge
  • Tension relief jaws, prevent tube-pulling, so less likelihood of operator error leaks
  • Motor operated electrode, with splash-guard-seals with single press of trigger
  • Single trigger activation minimizes repetitive strain injury risk
  • Available accessories to enhance efficiency
  • Communication port and data software for maintenance and use statistics
HWG2.00V includes a durable hard shell case that facilitates easy transport for fleet management while protecting device and peripherals.
Specifications
Voltage 100/240 V
For Use With (Application) Sealing Blood Bag Tubing
Includes Without Transport Case
Material Metal, Plastic