Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEN1/HEN2 Monoclonal Antibody (2A3)
GREENER_CHOICE

Catalog No. PIMA519463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519463 Supplier Invitrogen™ Supplier No. MA519463
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SDLEPVEEAE GDGKGGSRAA LYPHPQQLSR EEKRRRRRAT AKYRSAHATR ERIRVEAFN LAFAELRKLL PTLPPDKKLS KIEILRLAIC YISYLNHVLD V.

The helix-loop-helix (HLH) structures are known motifs commonly found in membrane-active and DNA-binding proteins. Helix-loop-helix proteins (HEN1 and HEN2) are DNA-binding proteins that may be involved in cell-type determination in the early nervous system. Studies of expression in normal tissues have demonstrated expression of NSCL-1 and NSCL-2 (genes encoding HEN1 and HEN2) in the developing central and peripheral nervous system, specifically in developing neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEN1/HEN2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2A3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NHLH2
Gene Accession No. Q02575, Q02577
Gene Alias 6230401I09Rik; bHLHa34; bHLHa35; Class A basic helix-loop-helix protein 34; class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; Hen1; HEN-1; HEN2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; NHLH1; Nhlh2; NSCL; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2
Gene Symbols NHLH1, NHLH2
Host Species Mouse
Immunogen NHLH2 (NP_005590, 36 a.a. ~ 135 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4807, 4808
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.