Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599778 Supplier Invitrogen™ Supplier No. PA599778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HEN2.

May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NHLH2
Gene Accession No. Q02577, Q64221
Gene Alias 6230401I09Rik; bHLHa34; bHLHa35; class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); helix-loop-helix protein 2; HEN1; HEN-1; Hen2; HEN-2; KIAA0490; Nescient helix loop helix 1; nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; Nhlh1; Nhlh2; Nscl; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2
Gene Symbols NHLH2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NHLH2(Accession Q02577), corresponding to amino acid residues A17-E67.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18072, 4808
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.