Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582607 Supplier Invitrogen™ Supplier No. PA582607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMLNSDTMEL DLPPTHSETE SGFSDCGGGA GPDGAGPGGP GGGQARGPEP GEPGRKDLQH LSREE Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 85%.

The helix-loop-helix (HLH) proteins are a family of putative transcription factors, some of which have been shown to play an important role in growth and development of a wide variety of tissues and species. Four members of this family have been clearly implicated in tumorigenesis via their involvement in chromosomal translocations in lymphoid tumors: MYC (MIM 190080), LYL1 (MIM 151440), E2A (MIM 147141), and SCL (MIM 187040).[supplied by OMIM, Nov 2002].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEN1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHLH1
Gene Accession No. Q02575
Gene Alias BHLHA35; class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); HEN1; HEN-1; nescient helix loop helix 1; nescient helix-loop-helix 1; Nhlh1; Nscl; NSCL1; NSCL-1; Tal2
Gene Symbols NHLH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NHLH1. Recombinant protein control fragment (Product #RP-92322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.