Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heneicosanoic acid, 99%

Catalog No. p-140374
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 2363-71-5 | C21H42O2 | 326.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2363-71-5
Molecular Formula C21H42O2
Molecular Weight (g/mol) 326.55
MDL Number MFCD00002805
InChI Key CKDDRHZIAZRDBW-UHFFFAOYSA-N
Synonym heneicosanoic acid, n-heneicosanoic acid, heneicosylic acid, henicosanoate, heneicosanate, heneicosanoate, n-heneicosanoate, heneicosanoicacid, heneicosanic acid, n-heneicosylic acid
PubChem CID 16898
ChEBI CHEBI:39248
IUPAC Name henicosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 74.0°C to 76.0°C
Color White
Boiling Point 170.0°C (0.1 mbar)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)19CO2H
Quantity 100 mg
Beilstein 02, I, 179
Solubility Information Solubility in water: .. Other solubilities: soluble in ether, hot acetone and hot hydrocarbons
Formula Weight 326.55
Percent Purity 99%
Physical Form Shiny Fluffy Crystalline Powder
Chemical Name or Material Heneicosanoic acid, 99%
Show More Show Less
EINECSNumber 219-113-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.