Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hentriacontane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014969
Click to view available options
Quantity:
250 mg
1 g

Hentriacontane is a normal alkane, comprising of approximately 31 carbon atoms, which can be found in beewax. It is also found to be a major component of candelilla wax, which upon organo-gelation can provide structure to solid-like fat-based foods, drug delivery systems and cosmetics.

Featured Industries: Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 630-04-6
Physical Form Neat
Linear Formula CH3(CH2)29 CH3
MDL Number MFCD00048745
Melting Point 67°C to 69°C
Type Analytical Standard
Quantity 1 g
Formula Weight 436.84
Percent Purity ≥98% (GC)
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.