Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hentriacontane 95.0+%, TCI America™
SDP

Catalog No. H001210G
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 630-04-6
Molecular Formula C31H64
Molecular Weight (g/mol) 436.85
MDL Number MFCD00048745
InChI Key IUJAMGNYPWYUPM-UHFFFAOYSA-N
Synonym n-hentriacontane, untriacontane, unii-6sdg640hl3, hentriacontan, candelilla wax, candellila wax, waxes, candelilla, triacontane, methyl, euphorbia cerifera wax, acmc-1b2c1
PubChem CID 12410
ChEBI CHEBI:5659
IUPAC Name hentriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 66°C
Color White
Boiling Point 180°C
Quantity 10 g
Formula Weight 436.85
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hentriacontane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.