Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Hep-Aid™ Disposable Kit

Catalog No. 19090609 Shop All Medique Products

Catalog No. 19-090-609

Supplier: Medique 86201

Includes

1 Clean-up Absorbent Pack, 1 Disposable Apron, 1 Disposable Goggles with Shields, 1 Pair Latex Gloves, 1 Scoop, Scraper, 2 Biohazard Bags, 1 8 oz bottle Chlorine concentrate, 3 Disposable Paper Towels, 1 Pair Shoe Covers, 1 Splash Protector Mask

Contains every thing to protect while cleaning and disposing of bodily fluids.

  • Kit includes clean-up absorbent pack, disposable apron, disposable goggles with shields, latex gloves, scoop, scraper, biohazard bags, 8 oz bottle chlorine concentrate, disposable paper towels, shoe covers, splash protector mask

Specifications

Product Type Hep-Aid™ Disposable Kit
For Use With (Application) Contains every thing to protect while cleaning and disposing of bodily fluids
Includes 1 Clean-up Absorbent Pack, 1 Disposable Apron, 1 Disposable Goggles with Shields, 1 Pair Latex Gloves, 1 Scoop, Scraper, 2 Biohazard Bags, 1 8 oz bottle Chlorine concentrate, 3 Disposable Paper Towels, 1 Pair Shoe Covers, 1 Splash Protector Mask
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.