Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEPACAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588157 Supplier Invitrogen™ Supplier No. PA588157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NITSPVRLIH GTVGKSALLS VQYSSTSSDR PVVKWQLKRD KPVTVVQSIG TEVIGTLRPD YRDRIRLFEN GSLLLSDLQL ADEGTYEVEI SITDDTFTGE KTINLTVDVP ISRPQVLVAS TTVLELSEAF TLNCSHENGT KPSYTWLKDG KPLLNDSRML LSPDQKVLTI TRVLMEDDDL YSCMVENPIS QGRSLPVKIT VYRRSSLYII L; Positive Samples: 293T, HepG2, MCF-7, SH-SY5Y, SGC-7901, Mouse brain, Mouse liver, Rat brain, Rat liver; Cellular Location: Cytoplasm, Cytoplasmic side, Membrane, Single-pass type I membrane protein.

HEPACAM is involved in regulating cell motility and cell-matrix interactions. HEPACAM may inhibit cell growth through suppression of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEPACAM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HEPACAM
Gene Accession No. Q14CZ8, Q640R3
Gene Alias 2900042E01Rik; GlialCAM; HEPACAM; hepatic and glial cell adhesion molecule; hepatocyte cell adhesion molecule; Hepn1; hypothetical protein FLJ25530; MLC2A; MLC2B; protein hepaCAM; RGD1306811
Gene Symbols HEPACAM
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-245 of human HEPACAM (NP_6899352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220296, 300517, 72927
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.