Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEPACAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597826
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597826 Supplier Invitrogen™ Supplier No. PA597826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEPACAM Polyclonal Antibody for Western Blot, ICC/IF, ELISA

HEPACAM is involved in regulating cell motility and cell-matrix interactions. HEPACAM may inhibit cell growth through suppression of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEPACAM
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HEPACAM
Gene Accession No. Q14CZ8
Gene Alias 2900042E01Rik; GlialCAM; HEPACAM; hepatic and glial cell adhesion molecule; hepatocyte cell adhesion molecule; Hepn1; hypothetical protein FLJ25530; MLC2A; MLC2B; protein hepaCAM; RGD1306811
Gene Symbols HEPACAM
Host Species Rabbit
Immunogen Recombinant Human Hepatocyte cell adhesion molecule protein (265-415AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220296
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.