Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEPACAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599999 Supplier Invitrogen™ Supplier No. PA599999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HEPACAM.

HEPACAM is involved in regulating cell motility and cell-matrix interactions. HEPACAM may inhibit cell growth through suppression of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEPACAM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HEPACAM
Gene Accession No. Q14CZ8, Q640R3
Gene Alias 2900042E01Rik; GlialCAM; HEPACAM; hepatic and glial cell adhesion molecule; hepatocyte cell adhesion molecule; Hepn1; hypothetical protein FLJ25530; MLC2A; MLC2B; protein hepaCAM; RGD1306811
Gene Symbols HEPACAM
Host Species Rabbit
Immunogen A synthesized peptide derived from human HEPACAM(Accession Q14CZ8), corresponding to amino acid residues S331-A381.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220296, 300517, 72927
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.