Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEPACAM2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281720
Description
HEPACAM2 Polyclonal specifically detects HEPACAM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
HEPACAM2 | |
Polyclonal | |
Unconjugated | |
PBS | |
HEPACAM2 | |
Rabbit polyclonal HEPACAM2 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human HEPACAM2. The immunogen is located within amino acids 370 - 420 of HEPACAM2. Amino Acid Squence: CVIKQKLEGRPETEYRKAQT | |
Affinity Purified | |
RUO | |
253012 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 0.5-1 μg/mL, ELISA | |
HEPACAM family member 2, MIKI | |
Rabbit | |
51 kDa | |
0.1 mg | |
Primary | |
HEPACAM2 antibody is human, mouse and rat reactive. At least two isoforms of HEPACAM2 are known to exist; this antibody will detect both isoforms. HEPACAM2 antibody is predicted to not cross-react with HEPACAM. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction