Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparanase-1, Rabbit anti-Human, Mouse, Polyclonal, Millipore Sigma™

Catalog No. ABC168725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC168725UG 25 μg
ABC1687100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC168725UG Supplier MilliporeSigma Supplier No. ABC168725UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Heparanase-1, Cat. No. ABC1687, is a rabbit polyclonal antibody that detects Heparanase-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Heparanase (UniProt: Q9Y251; also known as EC: 3.2.1.166, Endo-glucuronidase, Heparanase-1, Hpa1) is encoded by the HPSE (also known as HEP, HPA, HPA1, HPR1, HPSE1, HSE1) gene (Gene ID: 10855) in human. Heparanase is a heterodimeric, glycosylated, endoglycosidase that cleaves heparan sulfate proteoglycans (HSPGs) into heparan sulfate side chains and core proteoglycans and participates in extracellular matrix (ECM) degradation and remodeling. It is known to selectively cleave the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying either a 3-O-sulfo or a 6-O-sulfo group. It is also shown to cleave the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying a 2-O-sulfo group, but not linkages between a glucuronic acid unit and a 2-O-sulfated iduronic acid moiety. Heparanase is highly expressed in placenta and spleen and weakly expressed in lymph node, thymus, peripheral blood leukocytes, bone marrow, endothelial cells, fetal liver, and tumor tissues. Heparanase is synthesized with a signal peptide (aa 1-35) and a propeptide chain (110-157), which are cleaved off to produce the heterodimeric mature form of 8 kDa and 50 kDa subunits. This active 8/50 kDa heterodimer is resistant to degradation. Heparanase is essentially inactive at neutral pH but becomes active under acidic conditions such as during tumor invasion and in inflammatory processes. Heparanase activity is inhibited by EDTA and laminarian sulfate. Four isoforms of heparanase have been described that are produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heparanase-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Heparanase-1 in human spleen and human placenta tissue sections.Western Blotting Analysis: 1μg/mL from a representative lot detected Heparanase-1 in human spleen tissue lysate.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Endo-glucoronidase;Heparanase;Hpa1
Gene Symbols HPSE;HEP;HPA;HPA1;HPR1;HPSE1;HSE1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal half of larger subunit (50 kDa)of human Heparanase-1.
Molecular Weight of Antigen ∽59 kDa observed; 61.15 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001092010
Test Specificity This rabbit polyclonal antibody detects human Heparanase-1. It targets an epitope with in 13 amino acids from the C-terminal half of larger subunit (50 kDa).
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.