Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heparanase/HPSE Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314371
This item is not returnable.
View return policy
Description
Heparanase/HPSE Polyclonal antibody specifically detects Heparanase/HPSE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Heparanase/HPSE | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.2, endo-glucoronidase, HEP, heparanase, Heparanase-1, Hpa1, HPAheparanase exon 9 and 10 deletion, HPR1, HPSE1heparanase-1, HSE1heparanase exon10-deletion | |
Synthetic peptide | |
50 μg | |
Pancreatic Stem Cell Markers, Stem Cell Markers | |
10855 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction