Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Heparin Cofactor II Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA143055
Description
PA1-43055 detects SERPIND1 in human samples. PA1-43055 has successfully been used in Western blot and ELISA applications. PA1-43055 was derived from human heparin cofactor II.
The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency.
Specifications
Heparin Cofactor II | |
Polyclonal | |
Unconjugated | |
Serpind1 | |
AA985900; AI303446; D22S673; HC2; Hcf2; HCII; HC-II; Heparin cofactor 2; heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; LS2; Protease inhibitor leuserpin-2; rls2var1; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; serpin D1; serpin family D member 1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1; Serpind1; THPH10 | |
Sheep | |
SDS-PAGE | |
RUO | |
3053 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
8.9 mg/mL | |
50% water with 50% glycerol and no preservative | |
P05546 | |
Serpind1 | |
Human Heparin Cofactor II. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction