Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Heparin Cofactor II Recombinant Rabbit Monoclonal Antibody (25)
GREENER_CHOICE

Catalog No. PIMA529795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529795 Supplier Invitrogen™ Supplier No. MA529795
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse SerpinD1/HCII.

The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heparin Cofactor II
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 25
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Serpind1
Gene Accession No. P49182
Gene Alias AA985900; AI303446; D22S673; HC2; Hcf2; HCII; HC-II; Heparin cofactor 2; heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; LS2; Protease inhibitor leuserpin-2; rls2var1; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; serpin D1; serpin family D member 1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1; Serpind1; THPH10
Gene Symbols Serpind1
Host Species Rabbit
Immunogen Recombinant Mouse SerpinD1 protein (Met1-Ser478).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15160
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.