Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Heparin Monoclonal Antibody (T320.11)
GREENER_CHOICE

Catalog No. MA183771
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183771 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183771 Supplier Invitrogen™ Supplier No. MA183771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Heparin/Heparan sulfate, clone T320.11, recognizes the chemically related αβ-linked glycosaminoglycans heparin and heparan sulfate. Both heparin and heparan sulfate are composed of alternating sequences of glucosamine and uronic acid with heparin being the more heavily sulfated polymer (Gallagher & Walker, 1985).

Standard or unfractionated heparin is a complex mixture of naturally occurring glycosaminoglycans and is used as an anticoagulant to treat venous thrombosis or to prevent thrombosis in high risk patients. Heparin therapy is associated with frequent elevations in serum aminotransferase levels that are typically transient and not associated with clinical symptoms or significant liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=772].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heparin
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone T320.11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias Ardeparin; C26H42N2O37S5; Clexane; Enoxaparin; Lovenox; Nadroparin
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.