Learn More
Heparin sodium salt, from porcine intestinal mucosa, IU≥100/mg
Description
Heparin sodium salt is used as a blood anti-coagulant. It is also used as an effective inhibitor of mesenchymal cell proliferation. Further, it binds to antithrombin III, which inhibits coagulation proteases. It is used as a major component of serum free media for preventing cell aggregation. It is also used as an anti-cancer agent in in vitro cancer research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
- This compound can bind to antithrombin III and inhibit coagulation proteases
Applications
- In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
- Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
- It is also applied in laboratory studies related to stem cell differentiation and growth
- In in vitro studies, this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
Specifications
Specifications
| Quantity | 5 g |
| Merck Index | 14,4653 |
| Solubility Information | Soluble in water,dimethyl sulfoxide and ethanol. |
| Chemical Name or Material | Heparin Sodium Salt, from porcine intestinal mucosa |
Safety and Handling
| RTECSNumber | MI0850000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.