Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068095APC
Description
Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Hepatic Sinusoidal Endothelial Cells | |
Monoclonal | |
APC | |
Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells | |
Rat Hepatic Sinusoidal Endothelial Cells | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
SE-1 | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction