Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis A Surface Antigen Antibody (BGN/B5B3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Hepatitis A Surface Antigen |
|---|---|
| Clone | BGN/B5B3 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000 |
| Applications | Radioimmune Assays (RIA), ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017440
![]() |
Novus Biologicals
NB100621970.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10062197
![]() |
Novus Biologicals
NB10062197 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Hepatitis A Surface Antigen Monoclonal specifically detects Hepatitis A Surface Antigen in Virus samples. It is validated for ELISA, Radioimmunoassay.Specifications
| Hepatitis A Surface Antigen | |
| 1.0 mg/mL | |
| Radioimmune Assays (RIA), ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| IgG2a | |
| Protein G purified | |
| NB100-62197 is specific for Hepatitis A surface antigen. |
| BGN/B5B3 | |
| ELISA 1:100-1:2000, Radioimmunoassay 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lysate from foetal kidney cells of rhesus monkey infected with extract from patient with active viral Hepatitis A. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title