Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis A Virus Monoclonal Antibody (1889)
GREENER_CHOICE

Catalog No. PIMA533260
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533260 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533260 Supplier Thermo Scientific Supplier No. MA533260
Add to Cart
Add to Cart

Hepatitis A Virus Monoclonal Antibody (1889)

Hepatitis A Virus Monoclonal Antibody for ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis A Virus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1889
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias hepatitis a virus surface antigen; hepatitis a virus vp1; hepatitis a virus vp4
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.