Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis A Virus Vp3 Monoclonal Antibody (C22H)
GREENER_CHOICE

Catalog No. MA17371
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17371 Supplier Invitrogen™ Supplier No. MA17371
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7371 antibody reacts with Hepatitis A virus VP3 in viral and infected tissue samples. MA1-7371 has been successfully used in ELISA, Immunofluorescence and Immunohistochemical applications. The MA1-7371 antibody was raised against HAV VP3 protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis A Virus Vp3
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone C22H
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HAVgp1; HepA; hepatitis a virus surface antigen
Host Species Mouse
Immunogen VP3 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.