Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Antigen Antibody (3F6-G10), Novus Biologicals™
SDP

Catalog No. p-7201045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018253 0.025 mg
NB600727 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018253 Supplier Novus Biologicals Supplier No. NB6007270.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis B Antigen Monoclonal specifically detects Hepatitis B Antigen in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Hepatitis B Antigen
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3F6-G10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100
Formulation 0.1% Bovine Serum Albumin with 0.09% Sodium Azide
Gene Alias HBV X antigen, HBx, HBxAg
Host Species Mouse
Immunogen HB-Xag-Protein A Fusion protein
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.