Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Hepatitis B preS2 Antibody, PE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093580PE
Description
Hepatitis B preS2 Polyclonal antibody specifically detects Hepatitis B preS2 in Human samples. It is validated for Western BlotSpecifications
| Hepatitis B preS2 | |
| Polyclonal | |
| Western Blot | |
| HBV pre S2 Antigen, HBV pre S2 protein, Hepatitis B Virus pre S2 protein, PreS2/S antigen, S | |
| Synthetic peptide within residues 130-150 of the 42 kDa preS2 HBV protein. This sequence represents a B cell epitope. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction