Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis B Small S Protein Polyclonal Antibody

Catalog No. PIPA528812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528812 Supplier Invitrogen™ Supplier No. PA528812

Rabbit Polyclonal Antibody

Recommended positive controls: medium of 293T transfect HBV1a_small S (4 days). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The HBV surface protein antigens (HBsAg) are comprised of three carboxyl co terminal HBs proteins termed large (LHBs), middle (MHBs) and small (SHBs, also called major) protein. LHBs and MHBs also share the highly hydrophobic, repetitive, membrane spanning S domain. In addition, MHBs has a 55 amino acid region called preS2. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Small S Protein
Applications Western Blot
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Alias HBV; HBV1a; HBV1a small S; HepB; small S Protein
Host Species Rabbit
Immunogen Hepatitis B Virus 2a small S Protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.