Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B surface antigen Goat anti-Hepatitis B virus, Polyclonal, Abnova™

Catalog No. 89033695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-695 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-695 Supplier Abnova Corporation Supplier No. PAB29850
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg).

  • High specificity
  • High reactivity

Specifications

Antigen Hepatitis B surface antigen
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg).
Dilution ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Host Species Goat
Immunogen Native purified hepatitis B virus subtypes Ad and Ay.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Hepatitis B virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.