Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus core Ag Polyclonal Antibody, Abbomax

Catalog No. 502203633
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3633 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3633 Supplier Abbomax Inc Supplier No. 5001704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Hepatitis B Core Antigen Cellular location: Nuclei of infected cells.

Hepatitis B virus core Ag Polyclonal antibody specifically detects Hepatitis B virus core Ag in Virus samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Hepatitis B virus core Ag
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias ayw; HBc; HBcAg; HBV; HepB
Host Species Rabbit
Immunogen A synthetic peptide derived from internal sequence (31-178aa) of human hepatitis B Core Antigen
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.