Learn More
Invitrogen™ Hepatitis B Virus Core Antigen Monoclonal Antibody (H6F5)

Description
MA1-21697 detects Hepatitis B Virus Core Antigen in Virus samples.
Specifications
Specifications
| Antigen | Hepatitis B Virus Core Antigen |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | H6F5 |
| Concentration | 4.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | ayw; HBc; HBcAg; HBV; HepB |
| Host Species | Mouse |
| Immunogen | Recombinant HBeAg. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.