Learn More
Invitrogen™ Hepatitis B Virus E Monoclonal Antibody (1895)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547493
Description
Specificity: Specific for the e antigen of hepatitis B virus. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Hepatitis B e-antigen (HBeAg) is a viral protein associated with HBV infections. Unlike the surface antigen, the e-antigen is found in the blood only when there are viruses also present. When the virus goes into hiding, the e-antigen will no longer be present in the blood. HBeAg is often used as a marker of ability to spread the virus to other people (infectivity). Measurement of e-antigen may also be used to monitor the effectiveness of HBV treatment; successful treatment will usually eliminate HBeAg from the blood and lead to development of antibodies against e-antigen (anti-HBe). There are some types (strains) of HBV that do not make e-antigen; these are especially common in the Middle East and Asia. In areas where these strains of HBV are common, testing for HBeAg is not very useful.
Specifications
Hepatitis B Virus E | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA | |
1895 | |
Unconjugated | |
HBeAg; HBV; HepB | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.