Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis B Virus E Monoclonal Antibody (HBe7)

Catalog No. PIMA120273
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120273 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120273 Supplier Invitrogen™ Supplier No. MA120273

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MA1-20273 detects Hepatitis B virus E antigen in virus samples.

Hepatitis B e-antigen (HBeAg) is a viral protein associated with HBV infections. Unlike the surface antigen, the e-antigen is found in the blood only when there are viruses also present. When the virus goes into 'hiding,' the e-antigen will no longer be present in the blood. HBeAg is often used as a marker of ability to spread the virus to other people (infectivity). Measurement of e-antigen may also be used to monitor the effectiveness of HBV treatment; successful treatment will usually eliminate HBeAg from the blood and lead to development of antibodies against e-antigen (anti-HBe). There are some types (strains) of HBV that do not make e-antigen; these are especially common in the Middle East and Asia. In areas where these strains of HBV are common, testing for HBeAg is not very useful.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus E
Applications ELISA
Classification Monoclonal
Clone HBe7
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias HBeAg; HBV; HepB
Host Species Mouse
Immunogen Recombinant HBeAg.
Purification Method Ion-exchange chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.