Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Hepatitis B Virus E Monoclonal Antibody (J17H)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17376
Description
The MA1-7376 antibody reacts with Hepatitis B virus E antigen in viral and infected tissue samples. MA1-7376 has been successfully used in ELISA applications. The MA1-7376 antibody was raised against the HBV E antigen.
Hepatitis B e-antigen (HBeAg) is a viral protein associated with HBV infections. Unlike the surface antigen, the e-antigen is found in the blood only when there are viruses also present. When the virus goes into hiding, the e-antigen will no longer be present in the blood. HBeAg is often used as a marker of ability to spread the virus to other people (infectivity). Measurement of e-antigen may also be used to monitor the effectiveness of HBV treatment; successful treatment will usually eliminate HBeAg from the blood and lead to development of antibodies against e-antigen (anti-HBe). There are some types (strains) of HBV that do not make e-antigen; these are especially common in the Middle East and Asia. In areas where these strains of HBV are common, testing for HBeAg is not very useful.
Specifications
Hepatitis B Virus E | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA | |
J17H | |
Unconjugated | |
HBeAg; HBV; HepB | |
HBV E antigen. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction