Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis B Virus Surface Ad/Ay Polyclonal Antibody

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA173080

Catalog No. PA173080


Only null left
Add to Cart

Description

Description

The PA1-73080 antibody reacts with Hepatitis B surface antigen. PA1-73080 has been successfully used in ELISA applications. The PA1-73080 antibody was raised against the surface antigen subtypes ad & ay.

The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. The HBV surface protein antigens (HBsAg) are comprised of three carboxyl co terminal HBs proteins termed large (LHBs), middle (MHBs) and small (SHBs, also called major) protein. LHBs and MHBs also share the highly hydrophobic, repetitive, membrane spanning S domain. In addition, MHBs has a 55 amino acid region called preS2. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hepatitis B Virus Surface Ad/Ay
Polyclonal
Unconjugated
HBsAg; HBV; hepatitis b virus surface antigen; HepB
Surface antigen subtypes ad & ay.
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Goat
IgG Fraction
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.