Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus surface Ag Monoclonal Antibody (ABM136), Abbomax

Catalog No. 502203634
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3634 Supplier Abbomax Inc Supplier No. 5001714
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Hepatitis B virus surface Ag Cellular location: Infected cells.

Hepatitis B virus surface Ag Monoclonal antibody specifically detects Hepatitis B virus surface Ag in Virus samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Hepatitis B virus surface Ag
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ABM136
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Host Species Mouse
Immunogen A synthetic peptide derived from internal sequence ff human hepatitis B Surface Antigen
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.