Learn More
Invitrogen™ Hepatitis B Virus Surface Monoclonal Antibody (P3G9A8)

Description
MIH9705 targets Hepatitis B Surface Antigen in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MIH9705 immunogen is hepatitis B Surface Antigen. MIH9705 was formerly sold as a Seradyn product.
Specifications
Specifications
| Antigen | Hepatitis B Virus Surface |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | P3G9A8 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | HBsAg; HBV; hepatitis b virus surface antigen; HepB |
| Host Species | Mouse |
| Immunogen | Hepatitis B Surface Antigen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.