Learn More
Invitrogen™ Hepatitis B Virus Surface Monoclonal Antibody (P4A11)

Description
MA5-14731 targets Hepatitis B Surface Antigen in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MA5-14731 immunogen is hepatitis B Surface Antigen. Product MA514731 is a smaller package size of MIH9706 (formerly sold as a Seradyn product).
Specifications
Specifications
| Antigen | Hepatitis B Virus Surface |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | P4A11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene Alias | HBsAg; HBV; hepatitis b virus surface antigen; HepB |
| Host Species | Mouse |
| Immunogen | Hepatitis B Surface Antigen. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.