Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus X antigen Antibody (X36C), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222445R
Description
Hepatitis B Virus X antigen Monoclonal specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA.Specifications
Hepatitis B Virus X antigen | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HBVgp3 | |
Baculovirus expressed recombinant HBx. | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA | |
X36C | |
Western Blot, ELISA | |
HBVgp3 X protein HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein | |
Mouse | |
Protein A or G purified | |
RUO | |
944566 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction