Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Antibody (BGN/1246/5G7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Hepatitis C Virus |
---|---|
Clone | BGN/1246/5G7 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Western Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017698
![]() |
Novus Biologicals
NB100652510.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10065251
![]() |
Novus Biologicals
NB10065251 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Hepatitis C Virus Monoclonal specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Hepatitis C Virus | |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Virus | |
Synthetic peptide corresponding to the NS-5 antigen region of the native molecule. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
BGN/1246/5G7 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5 | |
IgG1 | |
Protein A purified | |
HEPATITIS C (BGN/1246/5G7) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title