Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | Hepatitis C Virus |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
Hepatitis C Virus Polyclonal specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot.Specifications
Hepatitis C Virus | |
Western Blot 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core, env, and part of E2/NS1 of HCV. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Virus | |
HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5 | |
IgG | |
Affinity Purified | |
Hepatitis C Virus (HCV) C3 protein |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title