Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus Core Antigen Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017929 0.025 mg
NB10093581 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017929 Supplier Novus Biologicals Supplier No. NB10093581SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis C Virus Core Antigen Polyclonal specifically detects Hepatitis C Virus Core Antigen in Human samples. It is validated for Western Blot.

Specifications

Antigen Hepatitis C Virus Core Antigen
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Core protein p19, HCcAg, HCV core antigen, HCV core protein
Gene Symbols POLY
Host Species Rabbit
Immunogen Hepatitis C Virus (HCV) core protein (HCV-C1)
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 951475
Test Specificity Hepatitis C Virus (HCV) core protein (HCV-C1)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.